CAS RN 81732-52-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bambuterol Monocarbamate Hydrochloride, TRC
CAS: 81732-52-7 Molecular Formula: C15 H24 N2 O4 . H Cl Molecular Weight (g/mol): 332.82 Synonym: Carbamic acid, dimethyl-, 3-[2-[(1,1-dimethylethyl)amino]-1-hydroxyethyl]-5-hydroxyphenyl ester, monohydrochloride (9CI),Bambuterol monocarbamate hydrochloride,3-[(1RS)-2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl]-5-hydroxyphenyl dimethylcarbamate hydrochloride IUPAC Name: [3-[2-(tert-butylamino)-1-hydroxyethyl]-5-hydroxyphenyl] N,N-dimethylcarbamate;hydrochloride SMILES: Cl.CN(C)C(=O)Oc1cc(O)cc(c1)C(O)CNC(C)(C)C