accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81777-89-1

H3CCH3ONClO

CAS RN 81777-89-1

IUPAC Name: 2-[(2-chlorophenyl)methyl]-4,4-dimethyl-1,2-oxazolidin-3-one
Synonyms: gamit command herbicide clomazone magister command dimethazone epa pesticide chemical code 125401 C12H14ClNO2
Molecular Weight (g/mol): 239.7
Molecular Formula: C12H14ClNO2
InChi Key: KIEDNEWSYUYDSN-UHFFFAOYSA-N
SMILES: CC1(C)CON(CC2=CC=CC=C2Cl)C1=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
  • (1)
Solvent or Matrix 
  • (1)
  • (2)
  • (2)
Concentration 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (1)
  • (1)
  • (3)
  • (5)
  • (3)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)