accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSnOCH3

CAS RN 818-08-6

IUPAC Name: dibutylstannanone
Synonyms: dibutyltin oxide
Molecular Weight (g/mol): 248.94
Molecular Formula: C8H18OSn
InChi Key: JGFBRKRYDCGYKD-UHFFFAOYSA-N
SMILES: CCCC[Sn](=O)CCCC
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (6)
  • (2)
  • (2)
  • (6)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)