accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 81885-67-8

CH3ONNH2

CAS RN 81885-67-8

IUPAC Name: 1-(7-amino-1,2,3,4-tetrahydroisoquinolin-2-yl)ethan-1-one
Synonyms: 1-(7-amino-3,4-dihydro-1H-isoquinolin-2-yl)ethanone
Molecular Weight (g/mol): 190.25
Molecular Formula: C11H14N2O
InChi Key: XWVFMJLNNGXNSG-UHFFFAOYSA-N
SMILES: CC(=O)N1CCC2=C(C1)C=C(N)C=C2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results