accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 819057-45-9

IUPAC Name: 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Synonyms: 3-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
Molecular Weight (g/mol): 237.08
Molecular Formula: C12H17BFNO2
InChi Key: FLMNWVXAEGUVNY-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C(F)C=C(N)C=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results