CAS RN 82-02-0
IUPAC Name:
4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one
Synonyms:
kelamin khellin ammi-khellin keloid kellin khellinum bi-kellina quelina khelline I chellina
Molecular Weight (g/mol):
260.25
Molecular Formula:
C14H12O5
InChi Key:
HSMPDPBYAYSOBC-UHFFFAOYSA-N
SMILES:
COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O
1
–
3
of
3
results
1
Thermo Scientific Chemicals Khellin, 95%
CAS: 82-02-0 Molecular Formula: C14H12O5 Molecular Weight (g/mol): 260.25 InChI Key: HSMPDPBYAYSOBC-UHFFFAOYSA-N IUPAC Name: 4,9-dimethoxy-7-methyl-5H-furo[3,2-g]chromen-5-one SMILES: COC1=C2OC=CC2=C(OC)C2=C1OC(C)=CC2=O