accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

O

CAS RN 82-05-3

IUPAC Name: tetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadeca-1(17),2,4,6,9,11,13,15-octaen-8-one
Synonyms: benzanthrone
Molecular Weight (g/mol): 230.27
Molecular Formula: C17H10O
InChi Key: HUKPVYBUJRAUAG-UHFFFAOYSA-N
SMILES: O=C1C2=CC=CC=C2C2=C3C(C=CC=C13)=CC=C2
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results