CAS RN 82-08-6
IUPAC Name:
(2E)-1-{6-[(3-acetyl-2,4,6-trihydroxy-5-methylphenyl)methyl]-5,7-dihydroxy-2,2-dimethyl-2H-chromen-8-yl}-3-phenylprop-2-en-1-one
Synonyms:
rottlerin kamalin mallotoxin
Molecular Weight (g/mol):
516.55
Molecular Formula:
C30H28O8
InChi Key:
DEZFNHCVIZBHBI-ZHACJKMWSA-N
SMILES:
CC(=O)C1=C(O)C(C)=C(O)C(CC2=C(O)C3=C(OC(C)(C)C=C3)C(C(=O)\C=C\C3=CC=CC=C3)=C2O)=C1O
1
–
3
of
3
results
2
Rottlerin, TRC
CAS: 82-08-6 Molecular Formula: C30 H28 O8 Molecular Weight (g/mol): 516.54 Synonym: 3'-((8-Cinnamoyl-5,7-dihydroxy-2,2-dimethyl-2H-1-benzopyran-6-yl)methyl)-2',4',6'-trihydroxy-5'-methyl-acetophenone,Mallotoxin IUPAC Name: (E)-1-[6-[(3-acetyl-2,4,6-trihydroxy-5-methyl-phenyl)methyl]-5,7-dihydroxy-2,2-dimethyl-chromen-8-yl]-3-phenyl-prop-2-en-1-one SMILES: CC(=O)c1c(O)c(C)c(O)c(Cc2c(O)c3C=CC(C)(C)Oc3c(C(=O)\C=C\c4ccccc4)c2O)c1O
3
R&D Systems™ Rottlerin
CAS: 82-08-6 Molecular Formula: C30H28O8