accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOHOO

CAS RN 82-75-7

IUPAC Name: 8-aminonaphthalene-1-sulfonic acid
Synonyms: 8-aminonaphthalene-1-sulfonic acid
Molecular Weight (g/mol): 223.25
Molecular Formula: C10H9NO3S
InChi Key: CYJJLCDCWVZEDZ-UHFFFAOYSA-N
SMILES: NC1=C2C(C=CC=C2S(O)(=O)=O)=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)