accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 821-48-7

IUPAC Name: bis(2-chloroethyl)azanium chloride
Synonyms: bis(2-chloroethyl)azanium chloride
Molecular Weight (g/mol): 178.48
Molecular Formula: C4H10Cl3N
InChi Key: YMDZDFSUDFLGMX-UHFFFAOYSA-N
SMILES: [Cl-].ClCC[NH2+]CCCl
Molecular Weight (g/mol) 
  • (3)
  • (4)
Percent Purity 
  • (3)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (4)
  • (3)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)