accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 822-39-9

IUPAC Name: 2-chloro-1,3,2-dioxaphospholane
Synonyms: 2-chloro-1,3,2-dioxaphospholane
Molecular Weight (g/mol): 126.48
Molecular Formula: C2H4ClO2P
InChi Key: OLSFRDLMFAOSIA-UHFFFAOYSA-N
SMILES: ClP1OCCO1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results