accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CNHNN

CAS RN 82241-22-3

IUPAC Name: 2-methyl-4-(piperazin-1-yl)quinoline
Synonyms: 2-methyl-4-(piperazin-1-yl)quinoline
Molecular Weight (g/mol): 227.31
Molecular Formula: C14H17N3
InChi Key: XYBLCORUTWKJOI-UHFFFAOYSA-N
SMILES: CC1=CC(N2CCNCC2)=C2C=CC=CC2=N1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results