accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OOO

CAS RN 823-82-5

IUPAC Name: furan-2,5-dicarbaldehyde
Synonyms: furan-2,5-dicarbaldehyde
Molecular Weight (g/mol): 124.1
Molecular Formula: C6H4O3
InChi Key: PXJJKVNIMAZHCB-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(O1)C=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results