accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HClH2NNHF

CAS RN 823-85-8

IUPAC Name: hydrogen (4-fluorophenyl)hydrazine chloride
Synonyms: hydrogen (4-fluorophenyl)hydrazine chloride
Molecular Weight (g/mol): 162.59
Molecular Formula: C6H8ClFN2
InChi Key: FEKUXLUOKFSMRO-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(F)C=C1
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (3)
  • (3)