accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 82385-42-0

NaOSNOO

CAS RN 82385-42-0

IUPAC Name: sodium 1,3-dioxo-3H-1λ⁶,2-benzothiazol-1-olate hydrate
Synonyms: sodium 1,3-dioxo-1λ⁶,2-benzothiazol-1-olate hydrate
Molecular Weight (g/mol): 223.18
Molecular Formula: C7H6NNaO4S
InChi Key: ZTTIVBTYHNUAIR-UHFFFAOYSA-M
SMILES: O.[Na+].[O-]S1(=O)=NC(=O)C2=CC=CC=C12
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (3)