accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

COOH

CAS RN 824-62-4

IUPAC Name: bicyclo[2.2.1]heptane-2-carboxylic acid
Synonyms: bicyclo[2.2.1]heptane-2-carboxylic acid
Molecular Weight (g/mol): 140.18
Molecular Formula: C8H12O2
InChi Key: JESWDXIHOJGWBP-UHFFFAOYNA-N
SMILES: OC(=O)C1CC2CCC1C2
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results