accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NOF

CAS RN 824-75-9

IUPAC Name: 4-fluorobenzamide
Synonyms: P-fluorobenzamide
Molecular Weight (g/mol): 139.13
Molecular Formula: C7H6FNO
InChi Key: VNDHYTGVCGVETQ-UHFFFAOYSA-N
SMILES: NC(=O)C1=CC=C(F)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)