accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOONa

CAS RN 824-79-3

IUPAC Name: sodium 4-methylbenzene-1-sulfinate
Synonyms: sodium 4-methylbenzenesulfinate
Molecular Weight (g/mol): 178.18
Molecular Formula: C7H7NaO2S
InChi Key: KFZUDNZQQCWGKF-UHFFFAOYSA-M
SMILES: [Na+].CC1=CC=C(C=C1)S([O-])=O
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)