accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NNONNOOHOHNH

CAS RN 82410-32-0

IUPAC Name: 2-amino-9-{[(1,3-dihydroxypropan-2-yl)oxy]methyl}-6,9-dihydro-3H-purin-6-one
Synonyms: ganciclovir biolf 62 monosodium salt ganciclovir biolf-62 nordeoxyguanosine dhpg sodium ganciclovir preparation cytovene-IV ganciclovir (as sodium) ganciclovir product
Molecular Weight (g/mol): 255.23
Molecular Formula: C9H13N5O4
InChi Key: IRSCQMHQWWYFCW-UHFFFAOYSA-N
SMILES: NC1=NC(=O)C2=C(N1)N(COC(CO)CO)C=N2
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (2)
  • (1)
Color 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results