accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHOHOHOHHOHOOHOH

CAS RN 82452-93-5

IUPAC Name: nonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21(53),22,24,27(52),28,30,33(51),34,36,39(50),40,42,45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Synonyms: nonacyclo[43.3.1.1³,⁷.1⁹,¹³.1¹⁵,¹⁹.1²¹,²⁵.1²⁷,³¹.1³³,³⁷.1³⁹,⁴³]hexapentaconta-1(49),3,5,7(56),9,11,13(55),15,17,19(54),21(53),22,24,27(52),28,30,33(51),34,36,39(50),40,42,45,47-tetracosaen-49,50,51,52,53,54,55,56-octol
Molecular Weight (g/mol): 848.99
Molecular Formula: C56H48O8
InChi Key: HDPRHRZFFPXZIL-UHFFFAOYSA-N
SMILES: OC1=C2CC3=CC=CC(CC4=CC=CC(CC5=CC=CC(CC6=C(O)C(CC7=C(O)C(CC8=C(O)C(CC9=C(O)C(CC1=CC=C2)=CC=C9)=CC=C8)=CC=C7)=CC=C6)=C5O)=C4O)=C3O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results