accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3(R)OHNNH3CCH3OHSOOO(R)(S)(R)(R)OH(S)(R)HO(S)CH3(S)(S)OH(R)CH3

CAS RN 82473-24-3

IUPAC Name: 3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylazaniumyl)-2-hydroxypropane-1-sulfonate
Synonyms: 3-({3-[(4R)-4-[(1R,3aS,3bR,4R,5aS,7R,9aS,9bS,11S,11aR)-4,7,11-trihydroxy-9a,11a-dimethyl-tetradecahydro-1H-cyclopenta[a]phenanthren-1-yl]pentanamido]propyl}dimethylammonio)-2-hydroxypropane-1-sulfonate
Molecular Weight (g/mol): 630.88
Molecular Formula: C32H58N2O8S
InChi Key: GUQQBLRVXOUDTN-XOHPMCGNSA-N
SMILES: C[C@H](CCC(=O)NCCC[N+](C)(C)CC(O)CS([O-])(=O)=O)[C@H]1CC[C@H]2[C@@H]3[C@H](O)C[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3C[C@H](O)[C@]12C
Form 
  • (2)
Product Type 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results