accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 82477-67-6

IUPAC Name: 3-chloro-5-methoxybenzoic acid
Synonyms: 3-chloro-5-methoxybenzoic acid
Molecular Weight (g/mol): 186.59
Molecular Formula: C8H7ClO3
InChi Key: RRVHCVOVXPMKGF-UHFFFAOYSA-N
SMILES: COC1=CC(Cl)=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results