accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OSO

CAS RN 825-44-5

IUPAC Name: 1λ⁶-benzothiophene-1,1-dione
Synonyms: benzo(B)thiophene S,S-dioxide
Molecular Weight (g/mol): 166.19
Molecular Formula: C8H6O2S
InChi Key: FRJNKYGTHPUSJR-UHFFFAOYSA-N
SMILES: O=S1(=O)C=CC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results