accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2SOOI

CAS RN 825-86-5

IUPAC Name: 4-iodobenzene-1-sulfonamide
Synonyms: 4-iodobenzenesulfonamide
Molecular Weight (g/mol): 283.08
Molecular Formula: C6H6INO2S
InChi Key: KQZFABSTXSNEQH-UHFFFAOYSA-N
SMILES: NS(=O)(=O)C1=CC=C(I)C=C1

 No Refinements