accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 825-99-0

IUPAC Name: 3-(methylsulfanyl)benzoic acid
Synonyms: 3-(methylsulfanyl)benzoic acid
Molecular Weight (g/mol): 168.21
Molecular Formula: C8H8O2S
InChi Key: PZGADOOBMVLBJE-UHFFFAOYSA-N
SMILES: CSC1=CC=CC(=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results