accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3NHNH2N

CAS RN 82560-12-1

IUPAC Name: 5-tert-butyl-1H-pyrazol-3-amine
Synonyms: 5-tert-butyl-1H-pyrazol-3-amine
Molecular Weight (g/mol): 139.2
Molecular Formula: C7H13N3
InChi Key: ZHBXGHWSVIBUCQ-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(N)=NN1
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results