accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3ONHOOHOH3CHCl

CAS RN 82586-62-7

IUPAC Name: 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Synonyms: 6,7-dimethoxy-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid hydrochloride
Molecular Weight (g/mol): 273.71
Molecular Formula: C12H16ClNO4
InChi Key: ROWPWZMWICGKBY-UHFFFAOYNA-N
SMILES: Cl.COC1=C(OC)C=C2CC(NCC2=C1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results