CAS RN 826-35-7
IUPAC Name:
1,2-dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Synonyms:
1,2-dimethyl (1R,2R)-cyclopropane-1,2-dicarboxylate
Molecular Weight (g/mol):
158.15
Molecular Formula:
C7H10O4
InChi Key:
JBVOSZYUSFDYIN-RFZPGFLSSA-N
SMILES:
COC(=O)[C@@H]1C[C@H]1C(=O)OC
1
–
4
of
4
results
2
Dimethyl trans-1,2-Cyclopropanedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Dimethyl trans-1,2-Cyclopropanedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Dimethyl trans-1,2-Cyclopropanedicarboxylate, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.