accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 826-81-3

H3COHN

CAS RN 826-81-3

IUPAC Name: 2-methylquinolin-8-ol
Synonyms: 2-methyl 8-quinolonol
Molecular Weight (g/mol): 159.19
Molecular Formula: C10H9NO
InChi Key: NBYLBWHHTUWMER-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=CC=C(O)C2=N1
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (6)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (2)