accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 827-99-6

IUPAC Name: 3-(trifluoromethoxy)phenol
Synonyms: 3-(trifluoromethoxy)phenol
Molecular Weight (g/mol): 178.11
Molecular Formula: C7H5F3O2
InChi Key: UWLJERQTLRORJN-UHFFFAOYSA-N
SMILES: OC1=CC=CC(OC(F)(F)F)=C1
Boiling Point 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (1)
  • (2)
  • (3)