accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H2NH3CBrCH3

CAS RN 82842-52-2

IUPAC Name: 3-bromo-2,4,6-trimethylaniline
Synonyms: 3-bromo-2,4,6-trimethylaniline
Molecular Weight (g/mol): 214.11
Molecular Formula: C9H12BrN
InChi Key: MVLMPTBHZPYDBZ-UHFFFAOYSA-N
SMILES: CC1=CC(C)=C(Br)C(C)=C1N
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results