accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 82846-18-2

IUPAC Name: 4-fluoro-3-methoxybenzoic acid
Synonyms: 4-fluoro-3-methoxybenzoic acid
Molecular Weight (g/mol): 170.14
Molecular Formula: C8H7FO3
InChi Key: LWGCZCMLPRMKIZ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=CC(=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results