accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3SOOSOOCl

CAS RN 82964-91-8

IUPAC Name: 4-methanesulfonylbenzene-1-sulfonyl chloride
Synonyms: 4-methanesulfonylbenzenesulfonyl chloride
Molecular Weight (g/mol): 254.7
Molecular Formula: C7H7ClO4S2
InChi Key: TYJOQICPGZGYDT-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)