accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COIOO

CAS RN 82998-57-0

IUPAC Name: 3-iodo-4-methoxybenzoate
Synonyms: 3-iodo-4-methoxybenzoate
Molecular Weight (g/mol): 277.04
Molecular Formula: C8H6IO3
InChi Key: HNJSYSWRPCCSQI-UHFFFAOYSA-M
SMILES: COC1=CC=C(C=C1I)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)