accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83-18-1

CH3OH3CN

CAS RN 83-18-1

IUPAC Name: 2,5-dimethyl-1-phenyl-1H-pyrrole-3-carbaldehyde
Synonyms: 2,5-dimethyl-1-phenylpyrrole-3-carbaldehyde
Molecular Weight (g/mol): 199.25
Molecular Formula: C13H13NO
InChi Key: LNROIXNEIZSESG-UHFFFAOYSA-N
SMILES: CC1=CC(C=O)=C(C)N1C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)