accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83-40-9

IUPAC Name: 2-hydroxy-3-methylbenzoic acid
Synonyms: cresotic acid acidum hydroxitoluicum hydroxytoluylsaeure 3-methylsalicylate β-cresotinate acido 3-ossi-5-metil-benzoico β-cresotinic acid 3-cresotinate cresotinic acid hydroxytoluic acid
Molecular Weight (g/mol): 152.15
Molecular Formula: C8H8O3
InChi Key: WHSXTWFYRGOBGO-UHFFFAOYSA-N
SMILES: CC1=CC=CC(C(O)=O)=C1O
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results