accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3C(S)(R)CH3(R)(S)(S)(S)OH(R)H3C(S)(R)H3CCH3H3C(E)

CAS RN 83-48-7

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-1-[(2R,3E,5S)-5-ethyl-6-methylhept-3-en-2-yl]-9a,11a-dimethyl-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: stigmasterol stigmasta-5,22-dien-3-b-ol phytosterol 豆甾醇 β-stigmasterol (24S)-5,22-stigmastadien-3b-ol stigmasterin stigmasta-5,22-dien-3β-ol stigmasta-5,22t-dien-3b-ol
Molecular Weight (g/mol): 412.7
Molecular Formula: C29H48O
InChi Key: HCXVJBMSMIARIN-PHZDYDNGSA-N
SMILES: CC[C@H](\C=C\[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C)C(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (4)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results