accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 830-13-7

O

CAS RN 830-13-7

IUPAC Name: cyclododecanone
Synonyms: cyclododecanone
Molecular Weight (g/mol): 182.31
Molecular Formula: C12H22O
InChi Key: SXVPOSFURRDKBO-UHFFFAOYSA-N
SMILES: O=C1CCCCCCCCCCC1
Molecular Weight (g/mol) 
  • (3)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)