accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 832-53-1

IUPAC Name: pentafluorobenzene-1-sulfonyl chloride
Synonyms: pentafluorobenzenesulfonyl chloride
Molecular Weight (g/mol): 266.57
Molecular Formula: C6ClF5O2S
InChi Key: UOJCTEGNHXRPKO-UHFFFAOYSA-N
SMILES: FC1=C(F)C(F)=C(C(F)=C1F)S(Cl)(=O)=O
Boiling Point 
  • (3)
  • (3)
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)