accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

BOHNCH3HOOH

CAS RN 832695-88-2

IUPAC Name: [3-(methylcarbamoyl)phenyl]boronic acid
Synonyms: 3-(methylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 178.98
Molecular Formula: C8H10BNO3
InChi Key: FYFFPNFUVMBPRZ-UHFFFAOYSA-N
SMILES: CNC(=O)C1=CC=CC(=C1)B(O)O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results