CAS RN 83280-65-3
IUPAC Name:
2-acetyl-4H,9H-naphtho[2,3-b]furan-4,9-dione
Synonyms:
2-acetylnaphtho[2,3-b]furan-4,9-dione
Molecular Weight (g/mol):
240.21
Molecular Formula:
C14H8O4
InChi Key:
DPHUWDIXHNQOSY-UHFFFAOYSA-N
SMILES:
CC(=O)C1=CC2=C(O1)C(=O)C1=CC=CC=C1C2=O
1
–
1
of
1
results
1
R&D Systems™ Napabucasin
A Tocris product, part of R&D Systems. STAT3 inhibitor; also blocks cancer stem cell self-renewal