accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83467-28-1

IUPAC Name: 4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
Synonyms: 4-(2-bromoethyl)-3,5-dimethyl-1H-pyrazole
Molecular Weight (g/mol): 203.08
Molecular Formula: C7H11BrN2
InChi Key: HGGLMDBNLJUEKA-UHFFFAOYSA-N
SMILES: CC1=C(CCBr)C(C)=NN1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results