accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83553-60-0

IUPAC Name: 5-(methylsulfanyl)-1H-imidazole
Synonyms: 4-(methylsulfanyl)-3H-imidazole
Molecular Weight (g/mol): 114.17
Molecular Formula: C4H6N2S
InChi Key: NLIJIXLRIYPOHM-UHFFFAOYSA-N
SMILES: CSC1=CN=CN1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results