accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83646-41-7

IUPAC Name: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5,7,9(13)-triene hydrobromide
Synonyms: julolidine hydrobromide
Molecular Weight (g/mol): 254.17
Molecular Formula: C12H16BrN
InChi Key: KHWBRFVBPGPEOJ-UHFFFAOYSA-N
SMILES: Br.C1CN2CCCC3=C2C(C1)=CC=C3
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results