accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83846-66-6

IUPAC Name: 1-(4-methylphenyl)cyclopropane-1-carboxylic acid
Synonyms: 1-(4-methylphenyl)cyclopropane-1-carboxylic acid
Molecular Weight (g/mol): 176.22
Molecular Formula: C11H12O2
InChi Key: AYUGAOYMYXSOKU-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)C1(CC1)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results