CAS RN 83846-86-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-Isopropylthioxanthone, TRC
CAS: 83846-86-0 Molecular Formula: C16 H14 O S Molecular Weight (g/mol): 254.35 Synonym: 4-(1-Methylethyl)-9H-thioxanthen-9-one,4-(1-Methylethyl)-9H-thioxanthen-9-one,4-Isopropyl-9-thioxanthone,4-Isopropyl-9H-thioxanthen-9-one,4-Isopropylthioxanthone IUPAC Name: 4-propan-2-ylthioxanthen-9-one SMILES: CC(C)c1cccc2C(=O)c3ccccc3Sc12