accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 83915-83-7

H2OH2OH2N(S)NH(S)OHOON(S)OHO

CAS RN 83915-83-7

IUPAC Name: (2S)-1-[(2S)-6-amino-2-{[(1S)-1-carboxy-3-phenylpropyl]amino}hexanoyl]pyrrolidine-2-carboxylic acid dihydrate
Synonyms: lisinopril dihydrate renacor
Molecular Weight (g/mol): 441.53
Molecular Formula: C21H35N3O7
InChi Key: CZRQXSDBMCMPNJ-ZUIPZQNBSA-N
SMILES: O.O.NCCCC[C@H](N[C@@H](CCC1=CC=CC=C1)C(O)=O)C(=O)N1CCC[C@H]1C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Certifications/Compliance 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results