accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3COBOH3CH3C(E)

CAS RN 83947-56-2

IUPAC Name: 4,4,5,5-tetramethyl-2-[(1E)-2-phenylethenyl]-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-[(1E)-2-phenylethenyl]-1,3,2-dioxaborolane
Molecular Weight (g/mol): 230.11
Molecular Formula: C14H19BO2
InChi Key: ARAINKADEARZLZ-ZHACJKMWSA-N
SMILES: CC1(C)OB(OC1(C)C)\C=C\C1=CC=CC=C1
Boiling Point 
  • (1)
  • (2)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)