accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHBrOHBr

CAS RN 84-59-3

IUPAC Name: 2,6-dibromonaphthalene-1,5-diol
Synonyms: 2,6-dibromonaphthalene-1,5-diol
Molecular Weight (g/mol): 317.96
Molecular Formula: C10H6Br2O2
InChi Key: GHJUWGHWJYULLK-UHFFFAOYSA-N
SMILES: OC1=C2C=CC(Br)=C(O)C2=CC=C1Br
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results