accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OOOOH3C

CAS RN 84-66-2

IUPAC Name: 1,2-diethyl benzene-1,2-dicarboxylate
Synonyms: diethyl phthalate diethyl o-phenylenediacetate unimoll DA palatinol A anozol neantine DEP diethyl o-phthalate phthalol placidol E
Molecular Weight (g/mol): 222.24
Molecular Formula: C12H14O4
InChi Key: FLKPEMZONWLCSK-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CC=CC=C1C(=O)OCC
Percent Purity 
  • (3)
  • (7)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (7)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (4)
  • (1)